Search
Search
| Gene Symbol: KDM4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096835.1 | XP_020952494.1 | 2857 | 78 | ||
| XM_021096836.1 | XP_020952495.1 | 2799 | 78 | |||
| GenBank mRNA | EU661937.1 | - | 1 - 2 | 67 | 78 | |
Set Membership: |
|