Search
Search
| Gene Symbol: LCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122329.3 | XP_003122377.2 | 306 | 60 | ||
| XM_021081121.1 | XP_020936780.1 | 313 | 60 | |||
| XM_021081122.1 | XP_020936781.1 | 312 | 60 | |||
| GenBank mRNA | FM995246.1 | - | 112 | 60 | ||
Set Membership: |
|