Search
Search
| Gene Symbol: ABCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655176.3 | XP_005655233.1 | 2516 | 72 | ||
| XM_013987836.2 | XP_013843290.1 | 2499 | 72 | |||
| XM_013987837.2 | XP_013843291.1 | 2422 | 72 | |||
| XM_021086765.1 | XP_020942424.1 | 2403 | 72 | |||
Set Membership: |
|