Search
Search

PHACTR2| Gene Symbol: PHACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087139.1 | XP_020942798.1 | 1825 | 91 | ||
| XM_021087148.1 | XP_020942807.1 | 1585 | 91 | |||
| XM_021087155.1 | XP_020942814.1 | 1608 | 91 | |||
| XM_021087161.1 | XP_020942820.1 | 1335 | 91 | |||
| XM_021087164.1 | XP_020942823.1 | 1416 | 91 | |||
Set Membership: |
|