Search
Search
| Gene Symbol: LRRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098586.1 | XP_020954245.1 | 3919 | 102 | ||
| XM_021098590.1 | XP_020954249.1 | 4096 | 102 | |||
| XM_021098593.1 | XP_020954252.1 | 2950 | 102 | |||
| XM_021098597.1 | XP_020954256.1 | 3711 | 102 | |||
Set Membership: |
|