Search
Search
| Gene Symbol: IL3RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK232356.1 | - | 1291 | 76 | ||
| AK346474.1 | - | 1291 | 76 | |||
| AK346543.1 | - | 1072 | 76 | |||
| AK396330.1 | - | 1295 | 76 | |||
| AK397812.1 | - | 1291 | 76 | |||
Set Membership: |
|