Search
Search
| Gene Symbol: GSDMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090504.1 | XP_020946163.1 | 1010 | 75 | ||
| XM_021090506.1 | XP_020946165.1 | 985 | 75 | |||
| GenBank mRNA | AK232388.1 | - | 6 - 7 | 922 | 75 | |
| AK232642.1 | - | 6 - 7 | 922 | 75 | ||
| AK394823.1 | - | 6 - 7 | 924 | 75 | ||
Set Membership: |
|