Search
Search
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124561.6 | XP_003124609.3 | 88 | |||
| XM_021086520.1 | XP_020942179.1 | 88 | ||||
| XM_021086521.1 | XP_020942180.1 | 88 | ||||
| GenBank mRNA | AK232522.1 | - | 5 - 6 | 830 | 88 | |
| AK393547.1 | - | 5 - 6 | 88 | |||
| AK393717.1 | - | 5 - 6 | 88 | |||
Set Membership: |
|