Search
Search
| Gene Symbol: NQO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100114.1 | XP_020955773.1 | 747 | 77 | ||
| XM_021100115.1 | XP_020955774.1 | 459 | 77 | |||
| XM_021100117.1 | XP_020955776.1 | 442 | 77 | |||
| XM_021100118.1 | XP_020955777.1 | 747 | 77 | |||
| XM_021100119.1 | XP_020955778.1 | 808 | 77 | |||
| GenBank mRNA | AK232559.1 | - | 4 - 5 | 614 | 77 | |
| AK234241.1 | - | 4 - 5 | 533 | 77 | ||
Set Membership: |
|