Search
Search
| Gene Symbol: TTLL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093096.1 | XP_020948755.1 | 2829 | 67 | ||
| GenBank mRNA | AK232777.1 | - | 1 - 2 | 52 | 67 | |
| AK398763.1 | - | 5 - 6 | 444 | 67 | ||
Set Membership: |
|