Search
Search
| Gene Symbol: KANK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083852.1 | XP_020939511.1 | 3170 | 60 | ||
| XM_021083853.1 | XP_020939512.1 | 3008 | 60 | |||
| XM_021083854.1 | XP_020939513.1 | 2991 | 60 | |||
| XM_021083855.1 | XP_020939514.1 | 3922 | 60 | |||
| XM_021083856.1 | XP_020939515.1 | 2700 | 60 | |||
| XM_021083857.1 | XP_020939516.1 | 2946 | 60 | |||
| XM_021083858.1 | XP_020939517.1 | 2831 | 60 | |||
| XM_021083859.1 | XP_020939518.1 | 2877 | 60 | |||
| XM_021083860.1 | XP_020939519.1 | 2578 | 60 | |||
| XM_021083861.1 | XP_020939520.1 | 2820 | 60 | |||
Set Membership: |
|