Search
Search
| Gene Symbol: AIPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981244.2 | XP_013836698.2 | 1212 | 70 | ||
| XM_013981245.2 | XP_013836699.2 | 1239 | 70 | |||
| XM_021067719.1 | XP_020923378.1 | 1166 | 70 | |||
Set Membership: |
|