Search
Search
| Gene Symbol: MEGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095212.1 | XP_020950871.1 | 4258 | 59 | ||
| XM_021095213.1 | XP_020950872.1 | 4153 | 59 | |||
| XM_021095214.1 | XP_020950873.1 | 4258 | 59 | |||
Set Membership: |
|