Search
Search
| Gene Symbol: MYBPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021085215.1 | XP_020940874.1 | 1738 | 97 | ||
| XM_021085216.1 | XP_020940875.1 | 1735 | 97 | |||
| XM_021085217.1 | XP_020940876.1 | 1699 | 97 | |||
| XM_021085218.1 | XP_020940877.1 | 1696 | 97 | |||
| XM_021085219.1 | XP_020940878.1 | 1738 | 97 | |||
| XM_021085220.1 | XP_020940879.1 | 1446 | 97 | |||