Search
Search
| Gene Symbol: TMCO6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124040.4 | XP_003124088.1 | 1013 | 83 | ||
| XM_013995233.2 | XP_013850687.1 | 1120 | 83 | |||
| XM_013995235.2 | XP_013850689.1 | 1086 | 83 | |||
| XM_013995236.2 | XP_013850690.1 | 1083 | 83 | |||
| XM_013995237.2 | XP_013850691.1 | 1047 | 83 | |||
| XM_013995238.2 | XP_013850692.1 | 970 | 83 | |||
| XM_013995239.2 | XP_013850693.1 | 1120 | 83 | |||
| XM_013995240.2 | XP_013850694.1 | 401 | 83 | |||
| XM_021084892.1 | XP_020940551.1 | 1013 | 83 | |||