Search
Search
| Gene Symbol: ZNF711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926232.7 | XP_001926267.5 | 1387 | 83 | ||
| XM_005673742.3 | XP_005673799.1 | 1383 | 83 | |||
| XM_005673744.3 | XP_005673801.1 | 1256 | 83 | |||
| XM_005673745.3 | XP_005673802.1 | 1125 | 83 | |||
| XM_005673746.3 | XP_005673803.1 | 998 | 83 | |||
| XM_021079909.1 | XP_020935568.1 | 1578 | 83 | |||
| XM_021079910.1 | XP_020935569.1 | 1602 | 83 | |||
| XM_021079911.1 | XP_020935570.1 | 1512 | 83 | |||
| XM_021079912.1 | XP_020935571.1 | 1378 | 83 | |||
| XM_021079913.1 | XP_020935572.1 | 1901 | 83 | |||
| XM_021079915.1 | XP_020935574.1 | 1383 | 83 | |||
| XM_021079916.1 | XP_020935575.1 | 1509 | 83 | |||
| XM_021079917.1 | XP_020935576.1 | 1252 | 83 | |||
| XM_021079918.1 | XP_020935577.1 | 1121 | 83 | |||
| XM_021079919.1 | XP_020935578.1 | 993 | 83 | |||
Set Membership: |
|