Search
Search
| Gene Symbol: TSEN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132372.6 | XP_003132420.3 | 1300 | 98 | ||
| XM_005669795.3 | XP_005669852.1 | 1248 | 98 | |||
| XM_013981990.2 | XP_013837444.1 | 1342 | 98 | |||
Set Membership: |
|