Search
Search
| Gene Symbol: PIBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021065658.1 | XP_020921317.1 | 1355 | 83 | ||
| XM_021065659.1 | XP_020921318.1 | 1206 | 83 | |||
| XM_021065660.1 | XP_020921319.1 | 1354 | 83 | |||
| XM_021065661.1 | XP_020921320.1 | 1205 | 83 | |||
| XM_021065663.1 | XP_020921322.1 | 1355 | 83 | |||
| XM_021065664.1 | XP_020921323.1 | 1355 | 83 | |||
| XM_021065665.1 | XP_020921324.1 | 1354 | 83 | |||
Set Membership: |
|