Search
Search
| Gene Symbol: THOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986394.2 | XP_013841848.1 | 2121 | 92 | ||
| XM_013986395.2 | XP_013841849.1 | 2121 | 92 | |||
| XM_013986396.2 | XP_013841850.1 | 2121 | 92 | |||
| XM_013986397.2 | XP_013841851.1 | 2119 | 92 | |||
| XM_021080460.1 | XP_020936119.1 | 2121 | 92 | |||
| XM_021080461.1 | XP_020936120.1 | 2121 | 92 | |||
| XM_021080462.1 | XP_020936121.1 | 2121 | 92 | |||
| XM_021080463.1 | XP_020936122.1 | 2121 | 92 | |||
| XM_021080464.1 | XP_020936123.1 | 2121 | 92 | |||
| XM_021080465.1 | XP_020936124.1 | 2119 | 92 | |||
| XM_021080466.1 | XP_020936125.1 | 2121 | 92 | |||
| XM_021080467.1 | XP_020936126.1 | 2121 | 92 | |||
| XM_021080468.1 | XP_020936127.1 | 2121 | 92 | |||
| XM_021080469.1 | XP_020936128.1 | 2119 | 92 | |||
| XM_021080470.1 | XP_020936129.1 | 2109 | 92 | |||
Set Membership: |
|