Search
Search
| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481931.4 | XP_003481979.2 | 222 | 96 | ||
| XM_005664951.3 | XP_005665008.2 | 222 | 96 | |||
| XM_005664952.3 | XP_005665009.2 | 222 | 96 | |||
| XM_013998924.2 | XP_013854378.2 | 222 | 96 | |||
Set Membership: |
|