Search
Search
| Gene Symbol: PHF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005673655.3 | XP_005673712.1 | 1597 | 65 | ||
| XM_005673656.3 | XP_005673713.2 | 2935 | 65 | |||
| XM_013986163.2 | XP_013841617.2 | 2036 | 65 | |||
| XM_021080703.1 | XP_020936362.1 | 1185 | 65 | |||
| XM_021080705.1 | XP_020936364.1 | 1600 | 65 | |||
Set Membership: |
|