Search
Search
| Gene Symbol: EHMT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081197.1 | XP_020936856.1 | 1277 | 79 | ||
| XM_021081198.1 | XP_020936857.1 | 1305 | 79 | |||
| XM_021081199.1 | XP_020936858.1 | 1256 | 79 | |||
| XM_021081200.1 | XP_020936859.1 | 1253 | 79 | |||
| XM_021081201.1 | XP_020936860.1 | 1277 | 79 | |||
| XM_021081202.1 | XP_020936861.1 | 1256 | 79 | |||