Search
Search
| Gene Symbol: TRIM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083408.1 | XP_020939067.1 | 1289 | 127 | ||
| XM_021083409.1 | XP_020939068.1 | 1045 | 127 | |||
| XM_021083410.1 | XP_020939069.1 | 528 | 127 | |||
| XM_021083411.1 | XP_020939070.1 | 608 | 127 | |||
Set Membership: |
|