Search
Search
| Gene Symbol: TMTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092123.1 | XP_020947782.1 | 2535 | 65 | ||
| XM_021092124.1 | XP_020947783.1 | 2136 | 65 | |||
| XM_021092125.1 | XP_020947784.1 | 2336 | 65 | |||