Search
Search

L3MBTL2| Gene Symbol: L3MBTL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929185.4 | XP_001929220.1 | 983 | 68 | ||
| XM_005653110.3 | XP_005653167.1 | 983 | 68 | |||
| XM_021091462.1 | XP_020947121.1 | 983 | 68 | |||