Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06931887_g1
See other PLXNB2 GE Assays ›
Gene Symbol:
PLXNB2
Gene Name:
plexin B2
Gene Aliases:
-
Chromosome Location:
Chr.5: 390347 - 416805 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06931887_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PLXNB2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021091277.1
XP_020946936.1
2847
74
XM_021091278.1
XP_020946937.1
3002
74
XM_021091279.1
XP_020946938.1
2967
74
XM_021091280.1
XP_020946939.1
2820
74
XM_021091281.1
XP_020946940.1
2820
74
XM_021091282.1
XP_020946941.1
2951
74
XM_021091283.1
XP_020946942.1
3115
74
XM_021091284.1
XP_020946943.1
2742
74
XM_021091286.1
XP_020946945.1
2937
74
XM_021091287.1
XP_020946946.1
2776
74
XM_021091288.1
XP_020946947.1
2919
74
XM_021091289.1
XP_020946948.1
2919
74
XM_021091290.1
XP_020946949.1
2999
74
XM_021091291.1
XP_020946950.1
2818
74
XM_021091292.1
XP_020946951.1
2934
74
XM_021091293.1
XP_020946952.1
3222
74
XM_021091294.1
XP_020946953.1
3114
74
XM_021091295.1
XP_020946954.1
2902
74
XM_021091296.1
XP_020946955.1
2741
74
XM_021091297.1
XP_020946956.1
2745
74
XM_021091298.1
XP_020946957.1
2844
74
XM_021091299.1
XP_020946958.1
2686
74
XM_021091300.1
XP_020946959.1
3041
74
XM_021091301.1
XP_020946960.1
2838
74
XM_021091302.1
XP_020946961.1
2818
74
XM_021091303.1
XP_020946962.1
2926
74
XM_021091304.1
XP_020946963.1
3222
74
XM_021091305.1
XP_020946964.1
2918
74
XM_021091307.1
XP_020946966.1
2748
74
XM_021091308.1
XP_020946967.1
2745
74
XM_021091309.1
XP_020946968.1
2590
74
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Sus scrofa
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline