Search
Search
| Gene Symbol: TLE6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021084149.1 | XP_020939808.1 | 1635 | 100 | ||
| XM_021084150.1 | XP_020939809.1 | 1861 | 100 | |||
| XM_021084151.1 | XP_020939810.1 | 1614 | 100 | |||
| XM_021084152.1 | XP_020939811.1 | 1593 | 100 | |||
| XM_021084153.1 | XP_020939812.1 | 1443 | 100 | |||
| XM_021084154.1 | XP_020939813.1 | 1466 | 100 | |||
| XM_021084155.1 | XP_020939814.1 | 1635 | 100 | |||
| XM_021084157.1 | XP_020939816.1 | 1359 | 100 | |||
Set Membership: |
|