Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06933239_g1
See other ADGRL2 GE Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
LPHN2
Chromosome Location:
Chr.6: 131727082 - 132366106 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06933239_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRL2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005656126.3
XP_005656183.1
2606
63
XM_005656127.3
XP_005656184.1
2563
63
XM_013988854.2
XP_013844308.1
2619
63
XM_013988855.2
XP_013844309.1
2565
63
XM_013988857.2
XP_013844311.1
2618
63
XM_013988862.2
XP_013844316.1
2564
63
XM_021096375.1
XP_020952034.1
3230
63
XM_021096376.1
XP_020952035.1
3055
63
XM_021096377.1
XP_020952036.1
3265
63
XM_021096378.1
XP_020952037.1
2600
63
XM_021096379.1
XP_020952038.1
3089
63
XM_021096380.1
XP_020952039.1
2924
63
XM_021096381.1
XP_020952040.1
2562
63
XM_021096382.1
XP_020952041.1
2477
63
XM_021096383.1
XP_020952042.1
2477
63
XM_021096384.1
XP_020952043.1
3089
63
XM_021096385.1
XP_020952044.1
3253
63
XM_021096386.1
XP_020952045.1
2594
63
XM_021096387.1
XP_020952046.1
3218
63
XM_021096388.1
XP_020952047.1
2578
63
XM_021096389.1
XP_020952048.1
2427
63
XM_021096390.1
XP_020952049.1
3184
63
XM_021096392.1
XP_020952051.1
2604
63
XM_021096393.1
XP_020952052.1
3241
63
XM_021096394.1
XP_020952053.1
2577
63
XM_021096395.1
XP_020952054.1
3259
63
XM_021096396.1
XP_020952055.1
2426
63
XM_021096397.1
XP_020952056.1
3212
63
XM_021096398.1
XP_020952057.1
2864
63
XM_021096399.1
XP_020952058.1
2445
63
XM_021096400.1
XP_020952059.1
2404
63
XM_021096402.1
XP_020952061.1
2867
63
XM_021096403.1
XP_020952062.1
2620
63
XM_021096404.1
XP_020952063.1
2620
63
XM_021096405.1
XP_020952064.1
2620
63
XM_021096406.1
XP_020952065.1
2619
63
XM_021096407.1
XP_020952066.1
2619
63
XM_021096408.1
XP_020952067.1
3225
63
XM_021096409.1
XP_020952068.1
2618
63
XM_021096410.1
XP_020952069.1
3226
63
XM_021096412.1
XP_020952071.1
2577
63
XM_021096413.1
XP_020952072.1
2564
63
XM_021096414.1
XP_020952073.1
3225
63
XM_021096415.1
XP_020952074.1
3186
63
XM_021096416.1
XP_020952075.1
2509
63
XM_021096417.1
XP_020952076.1
2428
63
XM_021096418.1
XP_020952077.1
2564
63
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Recommended Assay
Sus scrofa
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline