Search
Search
| Gene Symbol: FAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121666.6 | XP_003121714.3 | 2548 | 59 | ||
| XM_013993246.2 | XP_013848700.1 | 2453 | 59 | |||
| XM_013993247.2 | XP_013848701.1 | 2688 | 59 | |||
| XM_013993248.2 | XP_013848702.1 | 2527 | 59 | |||
| XM_013993249.2 | XP_013848703.1 | 2527 | 59 | |||
| XM_021098740.1 | XP_020954399.1 | 2527 | 59 | |||
| XM_021098743.1 | XP_020954402.1 | 2656 | 59 | |||
| XM_021098750.1 | XP_020954409.1 | 2521 | 59 | |||
| XM_021098755.1 | XP_020954414.1 | 2354 | 59 | |||
| XM_021098761.1 | XP_020954420.1 | 2527 | 59 | |||