Search
Search
| Gene Symbol: KMT2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079121.1 | XP_020934780.1 | 4648 | 69 | ||
| XM_021079122.1 | XP_020934781.1 | 4644 | 69 | |||
| XM_021079123.1 | XP_020934782.1 | 4621 | 69 | |||
| XM_021079124.1 | XP_020934783.1 | 4807 | 69 | |||
| XM_021079125.1 | XP_020934784.1 | 4807 | 69 | |||
| XM_021079126.1 | XP_020934785.1 | 4842 | 69 | |||
| XM_021079127.1 | XP_020934786.1 | 4807 | 69 | |||
| XM_021079128.1 | XP_020934787.1 | 4807 | 69 | |||
| XM_021079130.1 | XP_020934789.1 | 4759 | 69 | |||
| XM_021079131.1 | XP_020934790.1 | 4807 | 69 | |||
| XM_021079132.1 | XP_020934791.1 | 4807 | 69 | |||
| XM_021079133.1 | XP_020934792.1 | 4807 | 69 | |||
| XM_021079134.1 | XP_020934793.1 | 4807 | 69 | |||
| XM_021079135.1 | XP_020934794.1 | 4807 | 69 | |||
| XM_021079136.1 | XP_020934795.1 | 4807 | 69 | |||
| XM_021079137.1 | XP_020934796.1 | 4807 | 69 | |||
| XM_021079138.1 | XP_020934797.1 | 4807 | 69 | |||
| XM_021079140.1 | XP_020934799.1 | 4842 | 69 | |||