Search
Search
| Gene Symbol: MAPK11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091323.1 | XP_020946982.1 | 903 | 93 | ||
| XM_021091324.1 | XP_020946983.1 | 902 | 93 | |||
| XM_021091325.1 | XP_020946984.1 | 998 | 93 | |||
| XM_021091326.1 | XP_020946985.1 | 920 | 93 | |||
| XM_021091327.1 | XP_020946986.1 | 920 | 93 | |||
| XM_021091328.1 | XP_020946987.1 | 1816 | 93 | |||
| XM_021091329.1 | XP_020946988.1 | 1815 | 93 | |||
Set Membership: |
|