Search
Search
| Gene Symbol: MSI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001315780.1 | NP_001302709.1 | 8 - 9 | 538 | 131 | |
| XM_021066167.1 | XP_020921826.1 | 643 | 131 | |||
| XM_021066168.1 | XP_020921827.1 | 643 | 131 | |||
| XM_021066169.1 | XP_020921828.1 | 641 | 131 | |||
| XM_021066170.1 | XP_020921829.1 | 640 | 131 | |||
| XM_021066171.1 | XP_020921830.1 | 641 | 131 | |||
| XM_021066172.1 | XP_020921831.1 | 636 | 131 | |||
| XM_021066173.1 | XP_020921832.1 | 650 | 131 | |||
| XM_021066174.1 | XP_020921833.1 | 642 | 131 | |||
| XM_021066175.1 | XP_020921834.1 | 637 | 131 | |||
| XM_021066176.1 | XP_020921835.1 | 633 | 131 | |||
| XM_021066177.1 | XP_020921836.1 | 633 | 131 | |||
| GenBank mRNA | AB731109.2 | - | 8 - 9 | 538 | 131 | |
| AK395700.1 | - | 8 - 9 | 624 | 131 | ||
Set Membership: |
|