Search
Search
| Gene Symbol: RSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243642.1 | NP_001230571.1 | 2 - 3 | 732 | 117 | |
| XM_021093409.1 | XP_020949068.1 | 987 | 117 | |||
| XM_021093410.1 | XP_020949069.1 | 1057 | 117 | |||
| XM_021093411.1 | XP_020949070.1 | 1017 | 117 | |||
| XM_021093412.1 | XP_020949071.1 | 1081 | 117 | |||
| XM_021093413.1 | XP_020949072.1 | 927 | 117 | |||
| XM_021093414.1 | XP_020949073.1 | 1021 | 117 | |||
| XM_021093415.1 | XP_020949074.1 | 838 | 117 | |||
| GenBank mRNA | AK233814.1 | - | 2 - 3 | 732 | 117 | |
| AK234227.1 | - | 2 - 3 | 724 | 117 | ||
| AK234378.1 | - | 2 - 3 | 720 | 117 | ||
Set Membership: |
|