Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Vi06439657_s1
Gene Symbol:
Mumps virus strain Edinburgh 2
Gene Name:
Mumps rubulavirus
Gene Aliases:
Mumps virus strain Edinburgh 2, Mumps virus strain Edinburgh 4, Mumps virus strain Edinburgh 6, Mumps virus strain Matsuyama, Mumps virus strain Belfast, Mumps virus strain Enders, Mumps virus strain Jeryl Lynn, Mumps virus strain Kilham, Mumps virus Bristol 1, Mumps virus Miyahara vaccine, Mumps virus strain RW, Mumps virus strain SBL-1, Mumps virus strain Takahashi, Mumps virus strain Urabe vaccine AM9, Mumps virus genotype G, Mumps virus genotype F, Mumps virus genotype D, Mumps virus genotype H, Mumps virus genotype A, Mumps virus genotype B, Mumps virus genotype N, Mumps virus genotype C, Mumps virus genotype J, Mumps virus genotype K, Mumps virus genotype K/M, Mumps virus genotype G1, Mumps virus genotype G2, Mumps virus genotype H1, Mumps virus genotype H2, Mumps virus genotype D1, Mumps virus genotype I, Mumps virus genotype M, Mumps virus genotype L, Mumps orthorubulavirus, Mumps virus strain L-Zagreb, Mumps virus strain Urabe, Mumps virus strain SBL
Chromosome Location:
-
Species:
Mumps virus strain Edinburgh 2
Species Specific ID (Flybase ID):
-
Assay ID
Vi06439657_s1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Updated Mapping Information
This assay corresponds to CADT IDs APKA3DE and APYMJJD.
Set Membership:
Recommended Assay
Qualified
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline