Search
Search

olah| Gene Symbol: olah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001207.1 | NP_001001207.1 | 2 - 3 | 204 | 74 | |
| XM_012964486.2 | XP_012819940.1 | 2 - 3 | 607 | 74 | ||
| XM_018094603.1 | XP_017950092.1 | 2 - 3 | 211 | 74 | ||
| GenBank mRNA | BC067998.1 | - | 2 - 3 | 204 | 74 | |
| CR848638.2 | - | 2 - 3 | 164 | 74 | ||
Set Membership: |
|