Search
Search

chmp4b| Gene Symbol: chmp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004866.1 | NP_001004866.1 | 4 - 5 | 707 | 60 | |
| GenBank mRNA | BC074708.1 | - | 4 - 5 | 707 | 60 | |
| CR762287.2 | - | 4 - 5 | 674 | 60 | ||
Set Membership: |
|