Search
Search

c15orf40| Gene Symbol: c15orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004913.2 | NP_001004913.1 | 2 - 3 | 332 | 112 | |
| XM_012958825.2 | XP_012814279.1 | 3 - 4 | 698 | 112 | ||
| GenBank mRNA | BC075357.1 | - | 2 - 3 | 335 | 112 | |
Set Membership: |
|