Search
Search

rsph9| Gene Symbol: rsph9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005021.1 | NP_001005021.1 | 605 | 82 | ||
| GenBank mRNA | BC076699.1 | - | 605 | 82 | ||
| CR760050.2 | - | 532 | 82 | |||
Set Membership: |
|