Search
Search

gng2| Gene Symbol: gng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005038.1 | NP_001005038.1 | 3 - 4 | 204 | 94 | |
| XM_012968240.2 | XP_012823694.1 | 2 - 3 | 976 | 94 | ||
| XM_018096219.1 | XP_017951708.1 | 3 - 4 | 192 | 94 | ||
| XM_018096220.1 | XP_017951709.1 | 3 - 4 | 269 | 94 | ||
| GenBank mRNA | BC076900.1 | - | 3 - 4 | 204 | 94 | |
Set Membership: |
|