Search
Search

slc30a8| Gene Symbol: slc30a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011041.1 | NP_001011041.1 | 5 - 6 | 861 | 86 | |
| XM_018094625.1 | XP_017950114.1 | 4 - 5 | 870 | 86 | ||
| GenBank mRNA | BC084148.1 | - | 5 - 6 | 861 | 86 | |
Set Membership: |
|