Search
Search

psmb2| Gene Symbol: psmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011057.1 | NP_001011057.1 | 1 - 2 | 162 | 73 | |
| GenBank mRNA | BC084185.1 | - | 1 - 2 | 162 | 73 | |
| CR761567.2 | - | 1 - 2 | 155 | 73 | ||
Set Membership: |
|