Search
Search

pnhd| Gene Symbol: pnhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203534.1 | NP_988865.1 | 5 - 6 | 775 | 102 | |
| GenBank mRNA | AY333424.1 | - | 5 - 6 | 775 | 102 | |
| BC170803.1 | - | 5 - 6 | 766 | 102 | ||
| BC170805.1 | - | 5 - 6 | 766 | 102 | ||
| CR926370.2 | - | 5 - 6 | 751 | 102 | ||
Set Membership: |
|