Search
Search

cant1| Gene Symbol: cant1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203609.1 | NP_988940.1 | 2 - 3 | 813 | 61 | |
| XM_012971583.2 | XP_012827037.1 | 2 - 3 | 972 | 61 | ||
| XM_012971584.2 | XP_012827038.1 | 2 - 3 | 789 | 61 | ||
| XM_012971585.2 | XP_012827039.1 | 2 - 3 | 888 | 61 | ||
| GenBank mRNA | BC061377.1 | - | 2 - 3 | 813 | 61 | |
Set Membership: |
|