Search
Search

ppid| Gene Symbol: ppid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203653.1 | NP_988984.1 | 3 - 4 | 412 | 100 | |
| GenBank mRNA | BC061335.1 | - | 3 - 4 | 412 | 100 | |
| CT030612.1 | - | 3 - 4 | 353 | 100 | ||
Set Membership: |
|