Search
Search

atp1b2| Gene Symbol: atp1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203739.1 | NP_989070.1 | 6 - 7 | 859 | 72 | |
| GenBank mRNA | BC061283.1 | - | 6 - 7 | 875 | 72 | |
| CR760087.2 | - | 6 - 7 | 842 | 72 | ||
Set Membership: |
|