Search
Search

sag| Gene Symbol: sag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203742.1 | NP_989073.1 | 12 - 13 | 1140 | 83 | |
| XM_012970257.2 | XP_012825711.1 | 12 - 13 | 3066 | 83 | ||
| GenBank mRNA | BC061277.1 | - | 12 - 13 | 1140 | 83 | |
| CR760827.2 | - | 12 - 13 | 1075 | 83 | ||
Set Membership: |
|