Search
Search

qtrt1| Gene Symbol: qtrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203759.1 | NP_989090.1 | 8 - 9 | 1070 | 94 | |
| GenBank mRNA | BC062509.1 | - | 8 - 9 | 1070 | 94 | |
| CR760939.2 | - | 8 - 9 | 1069 | 94 | ||
| CR926355.1 | - | 8 - 9 | 1084 | 94 | ||
Set Membership: |
|