Search
Search

prdx6| Gene Symbol: prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203771.1 | NP_989102.1 | 2 - 3 | 261 | 86 | |
| GenBank mRNA | BC062510.1 | - | 2 - 3 | 261 | 86 | |
| BC169178.1 | - | 2 - 3 | 218 | 86 | ||
| CR760860.2 | - | 2 - 3 | 257 | 86 | ||
Set Membership: |
|