Search
Search

dnajb6| Gene Symbol: dnajb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203776.1 | NP_989107.1 | 6 - 7 | 637 | 106 | |
| XM_012964423.1 | XP_012819877.1 | 7 - 8 | 887 | 106 | ||
| XM_012964424.2 | XP_012819878.1 | 7 - 8 | 887 | 106 | ||
| GenBank mRNA | BC062492.1 | - | 6 - 7 | 637 | 106 | |
Set Membership: |
|