Search
Search

rps3| Gene Symbol: rps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203788.1 | NP_989119.1 | 4 - 5 | 382 | 86 | |
| GenBank mRNA | BC061265.1 | - | 4 - 5 | 382 | 86 | |
| CR848247.2 | - | 4 - 5 | 353 | 86 | ||
Set Membership: |
|